Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate 94%

Supplier: Thermo Scientific Chemicals
Danger

H26857-06 H26857-03 H26857-14 H26857-03
AAH26857-06EA 76.39 USD
AAH26857-06 AAH26857-03 AAH26857-14 CAAAH26857-03
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate 94%
Trimethylsilyl 2,2-difluoro-2-(fluorosulphonyl)acetate
Formula: C₅H₉F₃O₄SSi
MW: 250.27 g/mol
Boiling Pt: 156 °C
Density: 1.27
Flash Pt: 26 °C(79 °F)
Storage Temperature: Ambient
MDL Number: MFCD02093343
CAS Number: 120801-75-4
UN: 2920
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR