2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone 97%

Danger

Synonyms: 3,5-Bis(tert-butyl)-4-hydroxyphenacyl bromide2-Bromo-1-[3,5-di(tert-butyl)-4-hydroxyphenyl]ethan-1-one

H26835-03 H26835-06 H26835-14
AAH26835-03EA 78.62 USD
AAH26835-03 AAH26835-14 AAH26835-06
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone 97%
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
Formula: C₁₆H₂₃BrO₂
MW: 327.26 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00173889
CAS Number: 14386-64-2
UN: 1759
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR