4-Carboxyphenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
, 4-Carboxybenzeneboronic acid pinacol ester
H26825-06
H26825-03
AAH26825-03EA
35.17
USD
AAH26825-03
AAH26825-06
4-Carboxyphenylboronic acid pinacol ester 97%
4-Carboxyphenylboronic acid pinacol ester
Formula:
C₁₃H₁₇BO₄ MW: 248.08 g/mol Melting Pt: 227…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863710 CAS Number: 180516-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...