cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene

Supplier: Thermo Scientific Chemicals
Warning

H26815-03
AAH26815-03EA 167.99 USD
AAH26815-03
cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
Formula: C₁₀H₁₈N₄
MW: 194.28 g/mol
Melting Pt: 29…31 °C
Storage Temperature: Ambient
MDL Number: MFCD09263319
CAS Number: 79236-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR