(3-Formylindol-1-yl)acetic acid ≥99%
|
Warning
|
H26807-03
AAH26807-03EA
62.35
USD
AAH26807-03
(3-Formylindol-1-yl)acetic acid ≥99%
(3-Formylindol-1-yl)acetic acid
Formula:
C₁₁H₉NO₃ Melting Pt: ca 205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00452923 CAS Number: 138423-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...