2-Chloro-6-(trifluoromethyl)nicotinic acid 97%

Warning

H26805-ME
AAH26805-MEEA 100.86 USD
AAH26805-ME
2-Chloro-6-(trifluoromethyl)nicotinic acid 97%
2-Chloro-6-(trifluoromethyl)nicotinic acid
Formula: C₇H₃ClF₃NO₂
MW: 225.56 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD01862658
CAS Number: 280566-45-2
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR