2-Methoxy-4-(trifluoromethoxy)benzyl bromide 97%

Danger

H26771-03 H26771-06
AAH26771-03EA 124.5 USD
AAH26771-03 AAH26771-06
2-Methoxy-4-(trifluoromethoxy)benzyl bromide 97%
2-Methoxy-4-(trifluoromethoxy)benzyl bromide
Formula: C₉H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04115970
CAS Number: 886500-35-2
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR