Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate

Warning

Synonyms: 1,4,8,11-Tetraazacyclotetradecane-N,N',N'',N'''-tetraacetic acid, tetraethyl ester1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetic Acid Tetraethyl Ester

H26755-03 H26755-MD
AAH26755-MDEA 248.56 USD
AAH26755-MD AAH26755-03
Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate
Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate
Formula: C₂₆H₄₈N₄O₈
MW: 544.69 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD09263316
CAS Number: 126320-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR