2-Propionylfuran 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Furan-2-yl)propan-1-one

H26754-14 H26754-06
AAH26754-06EA 79.69 USD
AAH26754-06 AAH26754-14
2-Propionylfuran 97%
2-Propionylfuran
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 53…55 °C/1 mm
Melting Pt: 26…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00020936
CAS Number: 3194-15-8
EINECS: 221-693-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR