2,6-Dichloro-5-fluoronicotinonitrile 98%
|
Warning
|
Synonyms:
2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile
, 2,6-Dichloro-3-cyano-5-fluoropyridine
H26747-14
H26747-06
AAH26747-14EA
178.73
USD
AAH26747-14
AAH26747-06
2,6-Dichloro-5-fluoronicotinonitrile 98%
2,6-Dichloro-5-fluoronicotinonitrile
Formula:
C₆HCl₂FN₂ MW: 190.99 g/mol Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863289 CAS Number: 82671-02-1 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...