3-Chloro-2,4-difluorobenzylamine 97%

Danger

H26723-06 H26723-03
AAH26723-03EA 136.21 USD
AAH26723-03 AAH26723-06
3-Chloro-2,4-difluorobenzylamine 97%
3-Chloro-2,4-difluorobenzylamine
Formula: C₇H₆ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD06660335
CAS Number: 886761-65-5
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR