2-(tert-Butylsulfonyl)thioacetamide 97%

Warning

H26706-03 H26706-06
AAH26706-03EA 105.56 USD
AAH26706-03 AAH26706-06
2-(tert-Butylsulfonyl)thioacetamide 97%
2-(tert-Butylsulfonyl)thioacetamide
Formula: C₆H₁₃NO₂S₂
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00084920
CAS Number: 175277-31-3
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR