Dimethyl (1-ethylpropyl)malonate 97%

H26701-ME H26701-27
AAH26701-27EA 155.4 USD
AAH26701-27 AAH26701-ME
Dimethyl (1-ethylpropyl)malonate 97%
Dimethyl (1-ethylpropyl)malonate
Formula: C₁₀H₁₈O₄
Storage Temperature: Ambient
MDL Number: MFCD08436116
CAS Number: 39520-19-9
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR