Cyclohexanone cyanohydrin 98%

Danger

H26699-18 H26699-09
AAH26699-09EA 101.46 USD
AAH26699-09 AAH26699-18
Cyclohexanone cyanohydrin 98%
Cyclohexanone cyanohydrin
Formula: C₇H₁₁NO
Boiling Pt: 132 °C/19 mm
Melting Pt: 34…36 °C
Density: 1.031
Flash Pt: 118 °C(244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003818
EINECS: 213-246-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR