2,3-Difluoro-6-methoxybenzylamine 97%
|
Danger
|
H26670-03
H26670-MD
AAH26670-MDEA
54.11
USD
AAH26670-MD
AAH26670-03
2,3-Difluoro-6-methoxybenzylamine 97%
2,3-Difluoro-6-methoxybenzylamine
Formula:
C₈H₉F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD04116022 CAS Number: 886501-77-5 UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...