2-Amino-6-methoxy-3-nitropyridine 97%

Warning

Synonyms: 6-Methoxy-3-nitropyridin-2-amine

H26668-06
AAH26668-06EA 318.57 USD
AAH26668-06
2-Amino-6-methoxy-3-nitropyridine 97%
2-Amino-6-methoxy-3-nitropyridine
Formula: C₆H₇N₃O₃
MW: 169.14 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD03206481
CAS Number: 73896-36-3
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR