4-Fluoro-3-(trifluoromethoxy)benzonitrile 97%
|
Warning
|
H26667-06
H26667-03
AAH26667-06EA
509.49
USD
AAH26667-06
AAH26667-03
4-Fluoro-3-(trifluoromethoxy)benzonitrile 97%
4-Fluoro-3-(trifluoromethoxy)benzonitrile
Formula:
C₈H₃F₄NO Storage Temperature: Ambient |
MDL Number:
MFCD06660253 CAS Number: 886501-14-0 UN: 3276 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...