3-Chloro-5-fluoro-4-methoxybenzaldehyde 97%
|
Warning
|
H26666-03
AAH26666-03EA
113
USD
AAH26666-03
3-Chloro-5-fluoro-4-methoxybenzaldehyde 97%
3-Chloro-5-fluoro-4-methoxybenzaldehyde
Formula:
C₈H₆ClFO₂ Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115863 CAS Number: 177994-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...