3-Benzylamino-1-propanol 97%

Warning

Synonyms: N-(3-Hydroxypropyl)benzylamine3-(Benzylamino)-1-propanol3-(Benzylamino)propan-1-ol

H26664-MD H26664-03
AAH26664-03EA 235.72 USD
AAH26664-03 AAH26664-MD
3-Benzylamino-1-propanol 97%
3-Benzylamino-1-propanol
Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020622
CAS Number: 4720-29-0
EINECS: 225-210-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR