2,6-Difluoro-m-toluidine 97%

Warning

Synonyms: 2,6-Difluoro-3-methylaniline

H26656-06 H26656-03
AAH26656-06EA 429.7 USD
AAH26656-06 AAH26656-03
2,6-Difluoro-m-toluidine 97%
2,6-Difluoro-m-toluidine
Formula: C₇H₇F₂N
MW: 143.14
Storage Temperature: Ambient
MDL Number: MFCD06660182
CAS Number: 144851-63-8
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR