2-Fluoro-5-methylbenzylamine 97%
|
Danger
|
H26652-MD
H26652-03
AAH26652-03EA
143.62
USD
AAH26652-03
AAH26652-MD
2-Fluoro-5-methylbenzylamine 97%
2-Fluoro-5-methylbenzylamine
Formula:
C₈H₁₀FN Storage Temperature: Ambient |
MDL Number:
MFCD06213371 CAS Number: 93071-82-0 EINECS: None UN: 2735 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...