1-Boc-4-(hydroxymethyl)piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-BOC-4-Piperidinemethanol
, tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate
, 1-Boc-4-piperidinemethanol
H26650-03
H26650-06
AAH26650-03EA
33.07
USD
AAH26650-03
AAH26650-06
1-Boc-4-(hydroxymethyl)piperidine 97%
1-Boc-4-(hydroxymethyl)piperidine
Formula:
C₁₁H₂₁NO₃ MW: 215.29 g/mol Melting Pt: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD02094488 CAS Number: 123855-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...