Bis(toluene-4-sulfonyl)methane 97%

Supplier: Thermo Scientific Chemicals

H26617-06 H26617-MD H26617-03
AAH26617-06EA 353.99 USD
AAH26617-06 AAH26617-03 AAH26617-MD
Bis(toluene-4-sulfonyl)methane 97%
Bis(toluene-4-sulfonyl)methane
Formula: C₁₅H₁₆O₄S₂
MW: 324.42 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00032255
CAS Number: 15310-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR