3-(Ethoxycarbonyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 3-(Ethoxycarbonyl)benzeneboronic acid3-Ethoxycarbonylphenylboronic acid

H26615-06 H26615-03
AAH26615-03EA 69.7 USD
AAH26615-03 AAH26615-06
3-(Ethoxycarbonyl)phenylboronic acid 97%
3-(Ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
Melting Pt: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD02179444
CAS Number: 4334-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR