(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
AOP
, ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V)
, (7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate
, 7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate
, Tris(dimethylamino)(3H-1,2,3-triazolo[4,5-b]pyridin-3-yloxy)phosphonium Hexafluorophosphate
H26609-06
AAH26609-06EA
248.99
USD
AAH26609-06
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) 98+%
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )
Formula:
C₁₂H₂₂F₆N₆OP₂ MW: 443.27 g/mol Melting Pt: ca 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD09263289 CAS Number: 156311-85-2 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...