4-Methoxy-3-(trifluoromethyl)benzyl bromide 97%

Danger

H26606-03 H26606-06
AAH26606-03EA 125.43 USD
AAH26606-03 AAH26606-06
4-Methoxy-3-(trifluoromethyl)benzyl bromide 97%
4-Methoxy-3-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃O
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD01631578
CAS Number: 261951-89-7
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR