3-Fluoro-2-(trifluoromethyl)benzonitrile 97%
|
Warning
|
H26590-MD
H26590-03
AAH26590-MDEA
50.24
USD
AAH26590-MD
AAH26590-03
3-Fluoro-2-(trifluoromethyl)benzonitrile 97%
3-Fluoro-2-(trifluoromethyl)benzonitrile
Formula:
C₈H₃F₄N MW: 189.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631566 CAS Number: 261951-81-9 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...