Dimethyl (2-methylbutyl)malonate 97%

H26588-MD
AAH26588-MDEA 67.49 USD
AAH26588-MD
Dimethyl (2-methylbutyl)malonate 97%
Dimethyl (2-methylbutyl)malonate
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436117
CAS Number: 59107-99-2
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR