2,3-Dichloro-6-fluorobenzonitrile 97%

Warning

H26584-03 H26584-06
AAH26584-06EA 200.91 USD
AAH26584-06 AAH26584-03
2,3-Dichloro-6-fluorobenzonitrile 97%
2,3-Dichloro-6-fluorobenzonitrile
Formula: C₇H₂Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD04115878
CAS Number: 79544-26-6
EINECS: None
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR