(2,4-Dichlorophenoxy)thioacetamide 97%
|
Warning
|
Synonyms:
2-(2,4-Dichlorophenoxy)thioacetamide
H26583-03
H26583-MD
AAH26583-03EA
118.77
USD
AAH26583-03
AAH26583-MD
(2,4-Dichlorophenoxy)thioacetamide 97%
(2,4-Dichlorophenoxy)thioacetamide
Formula:
C₈H₇Cl₂NOS MW: 236.11 Melting Pt: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172128 CAS Number: 2302-32-1 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...