1,4-Benzodioxane-2-thiocarboxamide 97%
|
Warning
|
H26571-06
H26571-03
AAH26571-06EA
325.23
USD
AAH26571-06
AAH26571-03
1,4-Benzodioxane-2-thiocarboxamide 97%
1,4-Benzodioxane-2-thiocarboxamide
Formula:
C₉H₉NO₂S Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084889 CAS Number: 147031-85-4 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...