6-Chloro-2-fluoro-3-methylbenzonitrile 97%
|
Danger
|
H26558-03
H26558-09
AAH26558-03EA
71.93
USD
AAH26558-03
AAH26558-09
6-Chloro-2-fluoro-3-methylbenzonitrile 97%
6-Chloro-2-fluoro-3-methylbenzonitrile
Formula:
C₈H₅ClFN Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD06660186 CAS Number: 886502-19-8 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...