2-Phenylthioacetamide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26514-03
H26514-MD
AAH26514-MDEA
67.89
USD
AAH26514-MD
AAH26514-03
2-Phenylthioacetamide 97%
2-Phenylthioacetamide
Formula:
C₈H₉NS Melting Pt: 91…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022177 CAS Number: 645-54-5 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...