2-Valerylfuran 97%

Supplier: Thermo Scientific Chemicals
Warning

H26491-09 H26491-18
AAH26491-09EA 95.49 USD
AAH26491-09 AAH26491-18
2-Valerylfuran 97%
2-Valerylfuran
Formula: C₉H₁₂O₂
MW: 152.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053147
CAS Number: 3194-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR