Dimethyl (3,4-difluorobenzyl)malonate 97%

Warning

H26484-MD H26484-03
AAH26484-03EA 105.56 USD
AAH26484-03 AAH26484-MD
Dimethyl (3,4-difluorobenzyl)malonate 97%
Dimethyl (3,4-difluorobenzyl)malonate
Formula: C₁₂H₁₂F₂O₄
MW: 258.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07780242
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR