2-Phenoxythioacetamide 97%

Warning

H26478-MD
AAH26478-MDEA 55.24 USD
AAH26478-MD
2-Phenoxythioacetamide 97%
2-Phenoxythioacetamide
Formula: C₈H₉NOS
MW: 167.26 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00100035
CAS Number: 35370-80-0
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR