5-Methyl-2-(trifluoromethyl)benzyl bromide 97%
|
Danger
|
H26477-03
H26477-MD
AAH26477-MDEA
94.12
USD
AAH26477-MD
AAH26477-03
5-Methyl-2-(trifluoromethyl)benzyl bromide 97%
5-Methyl-2-(trifluoromethyl)benzyl bromide
Formula:
C₉H₈BrF₃ Storage Temperature: Ambient |
MDL Number:
MFCD06660296 CAS Number: 886502-86-9 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...