2-(4-Methoxyphenyl)piperazine 95%

Danger

H26476-MD
AAH26476-MDEA 400.71 USD
AAH26476-MD
2-(4-Methoxyphenyl)piperazine 95%
2-(4-Methoxyphenyl)piperazine
Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
Melting Pt: 55…63 °C
Storage Temperature: Ambient
MDL Number: MFCD05864677
CAS Number: 91517-26-9
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR