2-Fluoro-5-(trifluoromethoxy)benzonitrile 97%
|
Warning
|
H26458-09
H26458-03
AAH26458-03EA
52.1
USD
AAH26458-03
AAH26458-09
2-Fluoro-5-(trifluoromethoxy)benzonitrile 97%
2-Fluoro-5-(trifluoromethoxy)benzonitrile
Formula:
C₈H₃F₄NO MW: 205.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115894 CAS Number: 886498-08-4 EINECS: None UN: 3276 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...