4-Chloro-2,6-difluorobenzaldehyde 97%
|
Warning
|
H26455-09
AAH26455-09EA
936.5
USD
AAH26455-09
4-Chloro-2,6-difluorobenzaldehyde 97%
4-Chloro-2,6-difluorobenzaldehyde
Formula:
C₇H₃ClF₂O MW: 176.55 g/mol Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094460 CAS Number: 252004-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...