2,3-Difluoro-4-methoxybenzylamine 97%

Danger

H26452-MD H26452-03
AAH26452-MDEA 137.21 USD
AAH26452-MD AAH26452-03
2,3-Difluoro-4-methoxybenzylamine 97%
2,3-Difluoro-4-methoxybenzylamine
Formula: C₈H₉F₂NO
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD04115987
CAS Number: 886500-75-0
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR