4-Chloro-2,6-difluorobenzylamine 97%

Danger

H26446-03 H26446-06
AAH26446-03EA 138.78 USD
AAH26446-03 AAH26446-06
4-Chloro-2,6-difluorobenzylamine 97%
4-Chloro-2,6-difluorobenzylamine
Formula: C₇H₆ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD06660350
CAS Number: 518357-43-2
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR