2,6-Difluoro-4-methoxybenzylamine 97%

Danger

Synonyms: (2,6-Difluoro-4-methoxyphenyl)methanamine

H26432-03 H26432-06
AAH26432-03EA 159.84 USD
AAH26432-03 AAH26432-06
2,6-Difluoro-4-methoxybenzylamine 97%
2,6-Difluoro-4-methoxybenzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115924
CAS Number: 771573-20-7
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR