2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde 97%
|
Warning
|
Synonyms:
2,4-Bis(trifluoromethyl)-6-methoxybenzaldehyde
H26419-03
H26419-06
AAH26419-06EA
310.8
USD
AAH26419-06
AAH26419-03
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde 97%
2,5-Bis(trifluoromethyl)-6-methoxybenzaldehyde
Formula:
C₁₀H₆F₆O₂ MW: 272.15 Melting Pt: 36…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD06660316 CAS Number: 180134-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...