2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers) 97%

Warning

Synonyms: 2-Isobutyl-4,5-dimethyl-3-thiazoline4,5-Dimethyl-2-isobutyl-3-thiazoline

H26379-03
AAH26379-03EA 27.16 USD
AAH26379-03
2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers) 97%
2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers)
Formula: C₉H₁₇NS
MW: 171.30 g/mol
Boiling Pt: 208 °C
Density: 1.487
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD01680822
CAS Number: 65894-83-9
EINECS: 265-969-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR