(4-Fluoro-2-methylphenyl)acetic acid 97%
|
Warning
|
Synonyms:
2-(4-Fluoro-2-methylphenyl)acetic acid
H26365-03
H26365-MD
AAH26365-03EA
148.74
USD
AAH26365-03
AAH26365-MD
(4-Fluoro-2-methylphenyl)acetic acid 97%
(4-Fluoro-2-methylphenyl)acetic acid
Formula:
C₉H₉FO₂ MW: 168.17 g/mol Melting Pt: 96…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049318 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...