4-Morpholin-4-ylmethylbenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Morpholinomethyl)benzoic acid
H26364-03
AAH26364-03EA
165.99
USD
AAH26364-03
4-Morpholin-4-ylmethylbenzoic acid 97%
4-Morpholin-4-ylmethylbenzoic acid
Formula:
C₁₂H₁₅NO₃ MW: 221.26 g/mol Melting Pt: 179…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD01167722 CAS Number: 62642-62-0 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...