2-Fluoro-5-(trifluoromethoxy)benzylamine 97%

Danger

H26342-03 H26342-06
AAH26342-06EA 480.63 USD
AAH26342-06 AAH26342-03
2-Fluoro-5-(trifluoromethoxy)benzylamine 97%
2-Fluoro-5-(trifluoromethoxy)benzylamine
Formula: C₈H₇F₄NO
Storage Temperature: Ambient
MDL Number: MFCD04115895
CAS Number: 886498-13-1
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR