Ethyl-N-(2,4-dinitrophenoxy)acetimidate ≥98%

Warning

H26341-03
AAH26341-03EA 63.62 USD
AAH26341-03
Ethyl-N-(2,4-dinitrophenoxy)acetimidate ≥98%
Ethyl-N-(2,4-dinitrophenoxy)acetimidate
Formula: C₁₀H₁₁N₃O₆
MW: 269.21 g/mol
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00010133
CAS Number: 54322-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR