6-Chloro-2-fluoro-3-methylbenzyl bromide 97%

Danger

H26340-06 H26340-03
AAH26340-03EA 45.72 USD
AAH26340-03 AAH26340-06
6-Chloro-2-fluoro-3-methylbenzyl bromide 97%
6-Chloro-2-fluoro-3-methylbenzyl bromide
Formula: C₈H₇BrClF
Boiling Pt: 103 °C/3 mm
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD01631373
CAS Number: 261762-88-3
UN: 1759
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR